Proteintech
Manufacturer Code:110321AP150UL
Catalog # 50-172-6813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | alpha Actin Fusion Protein Ag1471 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTC ACTC1 ACTC1 cardiac muscle 1 Actin alpha cardiac muscle 1 actin alpha cardiac muscle 1 Alpha actin Alpha cardiac actin CMD1R CMH11; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; alpha-actin cardiac; Alpha-actin-1; Alpha-cardiac actin; alphac-actin; nemaline myopathy type 3
Gene Aliases: AA959943; ACTA; Acta-2; ACTA1; ACTC; Actc-1; ACTC1; Acts; Actsk-1; ASD5; ASMA; CFTD; CFTD1; CFTDM; CMD1R; CMH11; LVNC4; MPFD; NEM1; NEM2; NEM3; SHPM
UniProt ID: (Human) P68133, (Human) P68032, (Mouse) P68134, (Rat) P68136, (Mouse) P68033, (Rat) P68035
Entrez Gene ID: (Human) 58, (Human) 70, (Mouse) 11459, (Rat) 29437, (Mouse) 11464, (Rat) 29275
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.