R&D Systems
Manufacturer Code:BAM586
Catalog # BAM586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 129736 |
Immunogen | Rat urine-derived alpha 2u -Globulin |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 15.5 kDa FABP; 15.5 kDa fatty acid-binding protein; allergen n I; Allergen Rat n I; Alpha(2)-euglobulin; alpha-2u globulin PGCL1; Alpha-2u-globulin; Major urinary protein; MUP; Rat n 1
Gene Aliases: Mup
UniProt ID: (Rat) P02761
Entrez Gene ID: (Rat) 259246
Molecular Function: isomerase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.