Invitrogen
Manufacturer Code:MA527724
Catalog # PIMA527724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S330A-80 |
Immunogen | Synthetic peptide amino acids 442- 462 (QDFRRSFKHILFRRRRRGFRQ cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adra 2c Alpha-2CAR ADRA2C ADRA2L2 ADRARL2 Alpha 2CAR alpha2 AR C4 alpha2 C4 alpha2C ALPHA2CAR Alpha-2 adrenergic receptor subtype C4 Alpha-2C adrenoreceptor Adrenergic alpha 2C receptor Alpha 2C adrenergic receptor Alpha 2C adrenergic receptor subtype C4 Alpha 2C adrenoceptor Alpha 2C adrenoreceptor Alpha-2-adrenergic receptor renal type alpha2cii-adrenergic receptor Subtype alpha2 C4; adrenergic, alpha-2C-, receptor; Alpha-2 adrenergic receptor subtype C4; Alpha-2C adrenergic receptor; Alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha-2C-adrenergic receptor; alpha-2CAR; alpha2-AR-C4; subtype alpha2-C4
Gene Aliases: [a]2C; Adra-2c; ADRA2C; ADRA2L2; ADRA2RL2; ADRARL2; alpha2-C4; alpha2C; ALPHA2CAR
UniProt ID: (Human) P18825, (Mouse) Q01337
Entrez Gene ID: (Human) 152, (Mouse) 11553
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.