Invitrogen
Manufacturer Code:PA5114828
Catalog # PIPA5114828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Adrenergic Receptor alpha -2C corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [a]2C Adra 2c ADRA2C Adra-2c ADRA2L2 ADRA2RL2 ADRARL2 Adrenergic alpha2C- receptor class I adrenergic receptor alpha 2c adrenergic alpha-2C- receptor Adrenergic alpha2C- receptor class I adrenoceptor alpha 2C Alpha 2CAR alpha-2 adrenergic receptor subtype C4 alpha2 AR C4 alpha2 C4 alpha2-AR-C4 alpha2C Alpha-2C adrenergic receptor Alpha-2C adrenoceptor Alpha-2C adrenoreceptor alpha2-C4 alpha-2C-adrenergic receptor ALPHA2CAR alpha-2CAR Alpha-2C-AR subtype alpha2-C4; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; Alpha-2 adrenergic receptor subtype C4; Alpha-2C adrenergic receptor; Alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha-2C-adrenergic receptor; alpha-2CAR; alpha2-AR-C4; subtype alpha2-C4
Gene Aliases: [a]2C; Adra-2c; ADRA2C; ADRA2L2; ADRA2RL2; ADRARL2; alpha2-C4; alpha2C; ALPHA2CAR
UniProt ID: (Human) P18825, (Mouse) Q01337, (Rat) P22086
Entrez Gene ID: (Human) 152, (Mouse) 11553, (Rat) 24175
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.