Invitrogen
Manufacturer Code:PA577287
Catalog # PIPA577287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RPEPRGLPQ*SELNQE corresponding to amino acid residues 160-174 of rat alpha2B - Adrenoceptor with replacement of cysteine 169 (C169 ) with serine (*S) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2B; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; Alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; Alpha-2B adrenoreceptor; alpha-2BAR
Gene Aliases: Adra2b
UniProt ID: (Rat) P19328
Entrez Gene ID: (Rat) 24174
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.