Invitrogen
Manufacturer Code:MA112721
Catalog # 01-674-051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5G10 |
Immunogen | Synthetic peptide corresponding to the third intracellular loop of rat alpha 2b adrenergic receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; Alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; Alpha-2B adrenoreceptor; alpha-2BAR; subtype alpha2-C2
Gene Aliases: [a]2B; Adra-2b; Adra2b; alpha2-C2; alpha2B
UniProt ID: (Rat) P19328
Entrez Gene ID: (Mouse) 11552, (Rat) 24174
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.