Invitrogen
Manufacturer Code:PA572228
Catalog # PIPA572228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 223-256 amino acids from the Central region of DANRE adra2a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenergic, alpha-2A-, receptor; alpha(2A)AR; Alpha-2A adrenergic receptor Alpha-2A adrenoreceptor Alpha-2A adrenoceptor Alpha-2AAR adra2a; alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha-2AAR; alpha2A-adrenergic receptor
Entrez Gene ID: (Zebrafish) 266750
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.