Invitrogen
Manufacturer Code:PA518475
Catalog # PIPA518475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TERRPNGLGPERS) corresponding to the internal amino acids of ADRA2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA2A adrenergic alpha-2A- receptor ADRA2 ADRA2R ADRAR ALPHA2AAR ZNF32 OTTHUMP00000058849 alpha-2A-adrenergic receptor alpha-2AAR subtype C10; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR; alpha2A-adrenergic receptor; subtype alpha2-C10
Gene Aliases: Adra-2; Adra-2a; Adra2a; alpha(2A)AR; alpha2-C10; alpha2A; alpha2A-AR; AW122659
UniProt ID: (Mouse) Q01338
Entrez Gene ID: (Mouse) 11551
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.