Novus Biologicals
Manufacturer Code:NB10093554
Catalog # NB10093554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide comprising internal residues of the human Alpha 2c protein. Reacts with rat and mouse Alpha 2c. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA2L2alpha-2CAR ADRA2RL2 ADRARL2 adrenergic alpha-2C- receptor Alpha-2 adrenergic receptor subtype C4 alpha2-AR-C4 alpha-2C adrenergic receptor Alpha-2C adrenoceptor Alpha-2C adrenoreceptor alpha-2C-adrenergic receptor Alpha-2CAR ALPHA2CAR; adrenergic, alpha-2C-, receptor; Alpha-2 adrenergic receptor subtype C4; Alpha-2C adrenergic receptor; Alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha-2C-adrenergic receptor; alpha-2CAR; alpha2-AR-C4
Gene Aliases: ADRA2C; ADRA2L2; ADRA2RL2; ADRARL2; ALPHA2CAR
UniProt ID: (Human) P18825
Entrez Gene ID: (Human) 152
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.