R&D Systems
Manufacturer Code:FAB10324V100UG
Catalog # FAB10324V100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 491613 |
Immunogen | NS0 mouse myeloma cell line transfected with human alpha-2B Adrenergic R/ADRA2B Accession &num P18089 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a-2B AdrenergicR ADRA2B ADRA2B adrenergic alpha-2B- receptor ADRA2L1 ADRA2L1adrenergic receptor alpha 2B ADRA2RL1 ADRARL1 adrenergic alpha-2B- receptor Alpha-2 adrenergic receptor subtype C2 alpha-2-adrenergic receptor-like 1 alpha-2B adrenergic receptor alpha2B AdrenergicR Alpha-2B adrenoceptor Alpha-2B adrenoreceptor alpha-2B-adrenergic receptor Alpha-2BAR ALPHA2BAR G-protein coupled receptor; ADRA2B adrenergic, alpha-2B-, receptor; Alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; Alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; Alpha-2B adrenoreceptor; G-protein coupled receptor
Gene Aliases: ADRA2B; ADRA2L1; ADRA2RL1; ADRARL1; alpha-2BAR; ALPHA2BAR; FAME2
UniProt ID: (Human) P18089
Entrez Gene ID: (Human) 151
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.