Novus Biologicals
Manufacturer Code:NBP297424AF488
Catalog # NB101924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human alpha-2B Adrenergic R/ADRA2B fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA2B adrenergic alpha-2B- receptor ADRA2L1adrenergic receptor alpha 2B ADRA2RL1 ADRARL1 adrenergic alpha-2B- receptor Alpha-2 adrenergic receptor subtype C2 alpha-2-adrenergic receptor-like 1 alpha-2B adrenergic receptor Alpha-2B adrenoceptor Alpha-2B adrenoreceptor alpha-2B-adrenergic receptor Alpha-2BAR ALPHA2BAR G-protein coupled receptor; ADRA2B adrenergic, alpha-2B-, receptor; Alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; Alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; Alpha-2B adrenoreceptor; G-protein coupled receptor
Gene Aliases: ADRA2B; ADRA2L1; ADRA2RL1; ADRARL1; alpha-2BAR; ALPHA2BAR; FAME2
UniProt ID: (Human) P18089
Entrez Gene ID: (Human) 151
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.