R&D Systems
Manufacturer Code:FAB10129U100UG
Catalog # FB10129U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1006614 |
Immunogen | Synthetic peptide containing Human alpha-2A Adrenergic R/ADRA2A |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: a-2A AdrenergicR ADRA2 ADRA2A ADRA2Ralpha-2-adrenergic receptor platelet type ADRARZNF32 adrenergic alpha-2A- receptor Alpha-2 adrenergic receptor subtype C10 alpha-2A adrenergic receptor alpha2A AdrenergicR Alpha-2A adrenoceptor Alpha-2A adrenoreceptor Alpha-2AAR ALPHA2AAR alpha-2AAR subtype C10 ZNF32; adrenergic, alpha-2A-, receptor; Alpha-2 adrenergic receptor subtype C10; alpha-2-adrenergic receptor, platelet type; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR subtype C10
Gene Aliases: ADRA2; ADRA2A; ADRA2R; ADRAR; ALPHA2AAR; ZNF32
UniProt ID: (Human) P08913
Entrez Gene ID: (Human) 150
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.