R&D Systems
Manufacturer Code:MAB1938
Catalog # MAB1938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 257316 |
Immunogen | Human plasma-derived alpha 2 -Macroglobulin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A2M alpha 2Macroglobulin Alpha-2-M alpha-2-macroglobulin C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5 CPAMD5 CPAMD5DKFZp779B086 FWP007 S863-7; Alpha-2-M; Alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5
Gene Aliases: A2M; A2MD; CPAMD5; FWP007; S863-7
UniProt ID: (Human) P01023
Entrez Gene ID: (Human) 2
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.