Novus Biologicals
Manufacturer Code:NB300343
Catalog # NB300343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Alpha 2 Macroglobulin (sequence 1-1474) native human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2-M; alpha-2-M alpha-2-macroglobulin C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5 CPAMD5DKFZp779B086 FWP007 S863-7; Alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5
Gene Aliases: A2M; A2MD; CPAMD5; FWP007; S863-7
UniProt ID: (Human) P01023
Entrez Gene ID: (Human) 2
Molecular Function:
complement component
cytokine
defense/immunity protein
enzyme modulator
protease inhibitor
serine protease inhibitor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.