Invitrogen
Manufacturer Code:PA577286
Catalog # PIPA577286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EPVPPDERF*SGITEE corresponding to amino acid residues 231-245 of rat alpha1D -Adrenoceptor with replacement of cysteine 240 (C240 ) with serine (*S) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA1D ADRA1D; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; Alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; Alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; Alpha-adrenergic receptor 1a; RA42
Gene Aliases: [a]1d; Adra-1; ADRA1; ADRA1A; ADRA1D; ADRA1R; Adrd1; ALPHA1; alpha1D-AR; DAR; dJ779E11.2; Gpcr8; Spr8
UniProt ID: (Human) P25100, (Rat) P23944
Entrez Gene ID: (Human) 146, (Mouse) 11550, (Rat) 29413
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.