Invitrogen
Manufacturer Code:PA533329
Catalog # PIPA533329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 2nd extracellular domain of human ADRA1D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA1D Alpha 1d Alpha-1D adrenergic receptor ADRA1R Alpha-1D adrenoceptor Alpha-1D adrenoreceptor ALPHA1 Alpha-1D-adrenergic receptor Alpha-adrenergic receptor 1a Alpha1delta-AR DAR ADRA1 Adrenoceptor alpha 1D Alpha-1A adrenergic receptor DJ779E11.2; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; Alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; Alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; Alpha-adrenergic receptor 1a
Gene Aliases: ADRA1; ADRA1A; ADRA1D; ADRA1R; ALPHA1; DAR; dJ779E11.2
UniProt ID: (Human) P25100
Entrez Gene ID: (Human) 146
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.