Invitrogen
Manufacturer Code:PA532660
Catalog # PIPA532660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from C-terminal cytoplasmic domain of human ADRA1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA1 ALPHA1BAR adrenergic alpha-1B- receptor alpha-1B adrenergic receptor alpha-1B adrenoceptor alpha-1B adrenoreceptor alpha-1B-adrenergic receptor; adrenergic, alpha-1B-, receptor; Alpha-1B adrenergic receptor; Alpha-1B adrenoceptor; Alpha-1B adrenoreceptor; alpha-1B-adrenergic receptor
Gene Aliases: ADRA1; ADRA1B; ALPHA1BAR
UniProt ID: (Human) P35368
Entrez Gene ID: (Human) 147
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.