Invitrogen
Manufacturer Code:PA527120
Catalog # PIPA527120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 347 and 440 of alpha 1b Adrenergic Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA1 ALPHA1BAR alpha-1B adrenergic receptor alpha-1B adrenoceptor alpha-1B adrenoreceptor alpha-1B-adrenergic receptor; alpha-1B adrenergic receptor; alpha-1B adrenoceptor; alpha-1B adrenoreceptor; alpha1B-adrenergic receptor
Gene Aliases: [a]1b
Entrez Gene ID: (Mouse) 11548
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.