Invitrogen
Manufacturer Code:PA5114827
Catalog # PIPA5114827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Adrenergic Receptor alpha -1B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [a]1b ADA1B ADRA1 ADRA1B Adrenergic alpha 1B- receptor adrenergic receptor alpha 1b adrenergic alpha-1B- receptor adrenoceptor alpha 1B alpha 1B-adrenoceptor alpha 1B-adrenoreceptor Alpha-1B adrenergic receptor alpha-1B adrenoceptor Alpha-1B adrenoreceptor alpha-1B-adrenergic receptor alpha1B-adrenergic receptor ALPHA1BAR; Adrenergic alpha 1B- receptor; adrenergic receptor, alpha 1b; adrenergic, alpha-1B-, receptor; alpha 1B-adrenoceptor; alpha 1B-adrenoreceptor; Alpha-1B adrenergic receptor; Alpha-1B adrenoceptor; Alpha-1B adrenoreceptor; alpha-1B-adrenergic receptor; alpha1B-adrenergic receptor
Gene Aliases: [a]1b; ADRA1; ADRA1B; ALPHA1BAR
UniProt ID: (Human) P35368
Entrez Gene ID: (Human) 147, (Rat) 24173, (Mouse) 11548
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.