R&D Systems
Manufacturer Code:MAB10298100
Catalog # MAB10298100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 518508 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human alpha-1 Adrenergic R/ADRA1A Accession &num P35348 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a-1A AdrenergicR ADRA1A ADRA1Cadrenergic alpha-1A- receptor variant 1 ADRA1L1 adrenergic alpha -1A- receptor adrenergic alpha-1A- receptor adrenergic alpha-1A- receptor variant 3 adrenergic alpha-1A- receptor variant 5 adrenergic alpha-1A- receptor variant 8 adrenergic alpha-1C- receptor alpha-1A adrenergic receptor alpha1A AdrenergicR Alpha-1A adrenoceptor Alpha-1A adrenoreceptor ALPHA1AAR Alpha-1C adrenergic receptor Alpha-adrenergic receptor 1c G protein coupled receptor; adrenergic, alpha-1A-, receptor; Alpha-1A adrenergic receptor; Alpha-1A adrenoceptor; Alpha-1A adrenoreceptor; Alpha-1C adrenergic receptor; Alpha-adrenergic receptor 1c; G protein coupled receptor
Gene Aliases: ADRA1A; ADRA1C; ADRA1L1; ALPHA1AAR
UniProt ID: (Human) P35348
Entrez Gene ID: (Human) 148
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.