Invitrogen
Manufacturer Code:MA529032
Catalog # PIMA529032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11 |
Immunogen | Recombit Human Protein HC/Bikunin Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A1M EDC1 HCP HI30 IATIL ITI ITIL ITILC UTI; Alpha-1 microglycoprotein; Alpha-1-microglobulin; Bikunin; Complex-forming glycoprotein heterogeneous in charge; EDC1; growth-inhibiting protein 19; HI-30; Inter-alpha-trypsin inhibitor light chain; ITI-LC; Protein AMBP; Protein HC; Trypstatin; uristatin; Uronic-acid-rich protein
Gene Aliases: A1M; AMBP; EDC1; HCP; HI30; IATIL; ITI; ITIL; ITILC; UTI
UniProt ID: (Human) P02760
Entrez Gene ID: (Human) 259
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.