Invitrogen
Manufacturer Code:PA581584
Catalog # PIPA581584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Rat Protein HC/Bikunin/Ambp Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1M EDC1 HCP HI30 IATIL ITI ITIL ITILC UTI; alpha 1 microglobulin/bikunin; Alpha-1 microglobulin/bikunin; Alpha-1-microglobulin; Bikunin; HI-30; Inter-alpha-trypsin inhibitor light chain; ITI-LC; Protein AMBP; Trypstatin
Gene Aliases: Ambp; Itil
UniProt ID: (Rat) Q64240
Entrez Gene ID: (Rat) 25377
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.