R&D Systems
Manufacturer Code:AF7724SP
Catalog # AF7724SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human alpha 1-Microglobulin Gly20-Val203 predicted Accession # P02760 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: A1M alpha 1Microglobulin alpha-1-microglobulin/bikunin precursor AMBP bikunin EDC1 HCP HCPcomplex-forming glycoprotein heterogeneous in charge HI30 IATIL inter-alpha-trypsin inhibitor light chain ITI ITIL ITILC ITILgrowth-inhibiting protein 19 protein AMBP protein HC trypstatin uristatin uronic-acid-rich protein UTI; Alpha-1 microglycoprotein; Alpha-1-microglobulin; Bikunin; Complex-forming glycoprotein heterogeneous in charge; EDC1; growth-inhibiting protein 19; HI-30; Inter-alpha-trypsin inhibitor light chain; ITI-LC; Protein AMBP; Protein HC; Trypstatin; uristatin; Uronic-acid-rich protein
Gene Aliases: A1M; AMBP; EDC1; HCP; HI30; IATIL; ITI; ITIL; ITILC; UTI
UniProt ID: (Human) P02760
Entrez Gene ID: (Human) 259
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.