Abnova
Manufacturer Code:H00000223A01
Catalog # 89-004-011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALDH9A1 (NP_000687 173 a.a. ~ 270 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ALDH9A1.
For Research Use Only
Protein Aliases: 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family, member A1; Aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; ALDH4; ALDH7; ALDH9; E3; Gamma-aminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABA-DH; TMABADH
Gene Aliases: ALDH4; ALDH7; ALDH9; ALDH9A1; E3; TMABADH
UniProt ID: (Human) P49189
Entrez Gene ID: (Human) 223
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.