Abnova
Manufacturer Code:H00008659B01
Catalog # 89-006-610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALDH4A1 (NP_003739.2 1 a.a. ~ 563 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ALDH4A1 protein.
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 4 family, member A1; Aldehyde dehydrogenase family 4 member A1; ALDH4; Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase; P5CD; P5CDh; P5CDhL; P5CDhS
Gene Aliases: ALDH4; ALDH4A1; P5CD; P5CDH
UniProt ID: (Human) P30038
Entrez Gene ID: (Human) 8659
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.