Novus Biologicals
Manufacturer Code:NBP275398
Catalog # NBP275398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG82-31 |
Immunogen | Recombinant protein within human Alcohol Dehydrogenase aa 1-200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ADH alpha subunit ADH1alcohol dehydrogenase 1A alcohol dehydrogenase 1 (class I) alpha polypeptide alcohol dehydrogenase 1A (class I) alpha polypeptide Alcohol dehydrogenase subunit alpha aldehyde reductase EC 1.1.1 EC 1.1.1.1; ADH, alpha subunit; alcohol dehydrogenase 1 (class I), alpha polypeptide; Alcohol dehydrogenase 1A; Alcohol dehydrogenase subunit alpha; aldehyde reductase
Gene Aliases: ADH1; ADH1A
UniProt ID: (Human) P07327
Entrez Gene ID: (Human) 124
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.