Novus Biologicals
Manufacturer Code:NBP29282420UL
Catalog # NB027515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-375 of human ADH1C (NP_000660.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH gamma subunit ADH3alcohol dehydrogenase 1C alcohol dehydrogenase 1C (class I) gamma polypeptide alcohol dehydrogenase 3 (class I) gamma polypeptide Alcohol dehydrogenase subunit gamma aldehyde reductase EC 1.1.1 EC 1.1.1.1; ADH, gamma subunit; Alcohol dehydrogenase 1C; alcohol dehydrogenase 3 (class I), gamma polypeptide; Alcohol dehydrogenase subunit gamma; aldehyde reductase
Gene Aliases: ADH1C; ADH3
UniProt ID: (Human) P00326
Entrez Gene ID: (Human) 126
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.