Novus Biologicals
Manufacturer Code:NBP270092AF700
Catalog # N270092A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F12 |
Immunogen | Full length recombinant protein of human alcohol dehydrogenase 1B (NP_000659) produced in HEK293T cells. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ADH beta subunit ADH2alcohol dehydrogenase 1B alcohol dehydrogenase 1B (class I) beta polypeptide alcohol dehydrogenase 2 (class I) beta polypeptide Alcohol dehydrogenase subunit beta aldehyde reductase DKFZp686C06125 EC 1.1.1 EC 1.1.1.1; ADH, beta subunit; Alcohol dehydrogenase 1B; alcohol dehydrogenase 2 (class I), beta polypeptide; Alcohol dehydrogenase subunit beta; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117
Gene Aliases: ADH1B; ADH2; HEL-S-117
UniProt ID: (Human) P00325
Entrez Gene ID: (Human) 125
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.