Invitrogen
Manufacturer Code:PA529516
Catalog # PIPA529516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 461 and 723 of Human alanyl-tRNA synthetase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AARS; alanine tRNA ligase 1, cytoplasmic; Alanine--tRNA ligase, cytoplasmic; Alanyl-tRNA synthetase; alanyl-tRNA synthetase, cytoplasmic; AlaRS; CMT2N alaRS alanine tRNA ligase 1 cytoplasmic alanine--tRNA ligase cytoplasmic alanyl-tRNA synthetase cytoplasmic renal carcinoma antigen NY-REN-42; Protein sticky; Renal carcinoma antigen NY-REN-42; Sti
Gene Aliases: AARS; AARS1; AI316495; C76919; CMT2N; EIEE29; sti
UniProt ID: (Human) P49588, (Mouse) Q8BGQ7
Entrez Gene ID: (Human) 16, (Mouse) 234734
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.