Proteintech
Manufacturer Code:662001IG150UL
Catalog # 50-173-6633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7E5H8 |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acetylated alpha tubulin Acetylated tubulin Acetylated tubulin (Lys40) acetylated-a-tubulin acetyl-a-tubulin alpha tubulin Alpha tubulin 3 tubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 3; Alpha-tubulin isotype M-alpha-1; Detyrosinated tubulin alpha-1A chain; hum-a-tub1; hum-a-tub2; Talpha1 alpha-tubulin; tubulin alpha 3; Tubulin alpha-1 chain; Tubulin alpha-1A chain; Tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin, alpha 1; tubulin, alpha 1a; tubulin, alpha, brain-specific
Gene Aliases: B-ALPHA-1; LIS3; Tuba-1; Tuba1; TUBA1A; TUBA3
UniProt ID: (Human) Q71U36, (Mouse) P68369, (Rat) P68370
Entrez Gene ID: (Dog) 106557476, (Pig) 100135051, (Human) 7846, (Mouse) 22142, (Rat) 64158
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.