Abnova
Manufacturer Code:H00051099M01
Catalog # 89-014-309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F3 |
Immunogen | ABHD5 (AAH21958 1 a.a. ~ 349 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ABHD5.
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; Abhydrolase domain-containing protein 5; CDS; CGI58; IECN2; Lipid droplet-binding protein CGI-58; MGC8731; NCIE2; truncated abhydrolase domain-containing protein 5
Gene Aliases: ABHD5; CDS; CGI-58; CGI58; IECN2; NCIE2
UniProt ID: (Human) Q8WTS1
Entrez Gene ID: (Human) 51099
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.