Novus Biologicals
Manufacturer Code:NBP236768
Catalog # NBP236768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2502 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PAPKFSPVTPKFTPVASKFSPGAPGGSGSQPNQKLGHPEALSAGTGSPQPPSFTYAQQREKPRVQEKQHPVPPPAQNQNQVRSPGAPGPLTLKEVEELEQLTQQLMQDMEHPQRQNVAVNE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTCL tumor antigen se14-3 CTCL-associated antigen se14-3 cutaneous T-cell lymphoma associated antigen se14-3 Cutaneous T-cell lymphoma-associated antigen se14-3 KIAA1125 MGC31836 predicted protein of HQ2893 PRKCBP1 PRO2893 protein kinase C-binding protein 1 Rack7 RACK7protein kinase C binding protein 1 zinc finger MYND domain containing protein 8 Zinc finger MYND domain-containing protein 8 zinc finger MYND-type containing 8; Zyxin; Zyxin-2
Gene Aliases: ESP-2; HED-2; ZYX
UniProt ID: (Human) Q15942
Entrez Gene ID: (Human) 7791
Molecular Function: actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.