Proteintech
Manufacturer Code:602541IG150UL
Catalog # 50-173-6351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D7E7 |
Immunogen | Zyxin Fusion Protein Ag0419 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ESP 2 HED 2 ZYX zyxin Zyxin 2; Zyxin; Zyxin-2
Gene Aliases: 9530098H06Rik; ESP-2; HED-2; R75157; ZYX
UniProt ID: (Human) Q15942, (Mouse) Q62523
Entrez Gene ID: (Human) 7791, (Mouse) 22793
Molecular Function:
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.