Invitrogen
Manufacturer Code:PA519219
Catalog # PIPA519219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QQREKPRVQEKQH) corresponding to the internal amino acids of ZYX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ZYX zyxin ESP-2 HED-2; Zyxin; Zyxin-2
Gene Aliases: ESP-2; HED-2; ZYX
UniProt ID: (Human) Q15942
Entrez Gene ID: (Human) 7791
Molecular Function:
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.