Proteintech
Manufacturer Code:139661AP150UL
Catalog # 50-172-9125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZnT7 Fusion Protein Ag4957 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLC30A7 Zinc transporter 7 ZnT 7 Znt like transporter 2 ZNT7 ZnTL2; solute carrier family 30 (zinc transporter), member 7; Solute carrier family 30 member 7; Zinc transporter 7; zinc transporter like 2; zinc transporter ZnT-7; ZnT-7; Znt-like transporter 2
Gene Aliases: 1810059J10Rik; 2610034N15Rik; 4833428C12Rik; AI467242; SLC30A7; ZnT-7; ZNT7; ZNTL2
UniProt ID: (Human) Q8NEW0, (Mouse) Q9JKN1, (Rat) Q5BJM8
Entrez Gene ID: (Human) 148867, (Mouse) 66500, (Rat) 310801
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.