Proteintech
Manufacturer Code:173631AP150UL
Catalog # 50-173-1532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZnT3 Fusion Protein Ag10735 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLC30A3 Zinc transporter 3 ZnT 3 ZNT3; solute carrier family 30 (zinc transporter), member 3; Solute carrier family 30 member 3; solute carrier family 30 protein; Zinc transporter 3; zinc transporter ZnT-3; ZnT-3
Gene Aliases: SLC30A3; ZNT3
UniProt ID: (Human) Q99726, (Mouse) P97441, (Rat) Q6QIX3
Entrez Gene ID: (Human) 7781, (Mouse) 22784, (Rat) 366568
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.