Abnova
Manufacturer Code:H00007547M01
Catalog # 89-001-055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F7 |
Immunogen | ZIC3 (NP_003404.1 182 a.a. ~ 274 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ZIC3.
For Research Use Only
Protein Aliases: heterotaxy 1; HTX; HTX1; Zic family member 3 (odd-paired homolog, Drosophila); Zinc finger protein 203; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF203
Gene Aliases: HTX; HTX1; VACTERLX; ZIC3; ZNF203
UniProt ID: (Human) O60481
Entrez Gene ID: (Human) 7547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.