Novus Biologicals
Manufacturer Code:NBP297488IR
Catalog # NB044158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Zeta Opioid Receptor fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7-60 7-60 protein OGFr opioid growth factor receptor Protein 7-60 Zeta-type opioid receptor; OGFr; Opioid growth factor receptor; Protein 7-60; Zeta-type opioid receptor
Gene Aliases: OGFR
UniProt ID: (Human) Q9NZT2
Entrez Gene ID: (Human) 11054
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.