Invitrogen
Manufacturer Code:PA5101968
Catalog # PIPA5101968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ZZZ3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3110065C23Rik 6430567E01Rik AA408566 AI481057 ATAC component 1 homolog ATAC1 AW556059 DKFZP564I052 R75514 RGD1565468 zinc finger ZZ-type containing 3 zinc finger ZZ domain containing 3 zinc finger ZZ-type containing 3 ZZ-type zinc finger-containing protein 3 Zzz3; ATAC component 1 homolog; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3
Gene Aliases: 3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC1; AW556059; R75514; RGD1565468; ZZZ3
UniProt ID: (Human) Q8IYH5, (Mouse) Q6KAQ7
Entrez Gene ID: (Human) 26009, (Mouse) 108946, (Rat) 310958
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.