Proteintech
Manufacturer Code:226081AP150UL
Catalog # 50-173-3904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZZZ3 Fusion Protein Ag18362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATAC component 1 homolog; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3; ZZZ3
Gene Aliases: ATAC1; ZZZ3
UniProt ID: (Human) Q8IYH5
Entrez Gene ID: (Human) 26009
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.