Abnova
Manufacturer Code:H00007791A01
Catalog # 89-105-776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 572 a.a) full-length recombinant protein with GST tag.,ZYX (AAH08743 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant ZYX.
For Research Use Only
Protein Aliases: ESP-2; HED-2; Zyxin; Zyxin-2
Gene Aliases: ESP-2; HED-2; ZYX
UniProt ID: (Human) Q15942
Entrez Gene ID: (Human) 7791
Molecular Function:
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.