Invitrogen
Manufacturer Code:PA543570
Catalog # PIPA543570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZXDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SERH2790; Zinc finger protein ZXDC; ZXD family zinc finger C; ZXD family zinc finger C-like; ZXD-like zinc finger protein
Gene Aliases: A930012H20; B930086F11Rik; BC003332; RGD1561296; ZXDC; ZXDL
UniProt ID: (Human) Q2QGD7, (Mouse) Q8C8V1
Entrez Gene ID: (Horse) 100060787, (Human) 79364, (Dog) 609097, (Rabbit) 100358258, (Mouse) 80292, (Rat) 362399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.