Proteintech
Manufacturer Code:205301AP150UL
Catalog # 50-173-2940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZXDC Fusion Protein Ag14392 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SERH2790; Zinc finger protein ZXDC; Zinc finger protein ZXDC ZXD family zinc finger C ZXD like zinc finger protein ZXDC ZXDL; ZXD-like zinc finger protein
Gene Aliases: A930012H20; B930086F11Rik; BC003332; ZXDC; ZXDL
UniProt ID: (Human) Q2QGD7, (Mouse) Q8C8V1
Entrez Gene ID: (Human) 79364, (Mouse) 80292
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.