Invitrogen
Manufacturer Code:MA537552
Catalog # PIMA537552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 422CT4.6.4 |
Immunogen | purified His-tagged ZWINT protein (fragment) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2010007E07Rik 2600001N01Rik D10Ertd749e human ZW10 interacting protein-1 HZwint-1 KNTC2AP neuroprotective protein 8 scoilin Sip30 SNAP25 interacting protein 30 SNAP25 interacting protein of 30 kDa SNAP25-interacting protein 30 ZW10 interacting kinetochore protein ZW10 interacting protein-1 ZW10 interactor ZW10 interactor kinetochore protein ZW10-interacting protein 1 Zwint ZWINT1 Zwint-1; human ZW10 interacting protein-1; SNAP25 interacting protein of 30 kDa; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; Zwint-1
Gene Aliases: 2010007E07Rik; 2600001N01Rik; D10Ertd749e; HZwint-1; KNTC2AP; SIP30; ZWINT; Zwint-1; ZWINT1
UniProt ID: (Human) O95229, (Mouse) Q9CQU5
Entrez Gene ID: (Human) 11130, (Mouse) 52696
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.