Invitrogen
Manufacturer Code:PA592903
Catalog # PIPA592903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-277 of human ZWINT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: human ZW10 interacting protein-1; neuroprotective protein 8; Scoilin; SNAP25 interacting protein 30; SNAP25 interacting protein of 30 kDa; SNAP25-interacting protein 30; ZW10 interacting protein-1; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; Zwint-1
Gene Aliases: 2010007E07Rik; 2600001N01Rik; D10Ertd749e; HZwint-1; KNTC2AP; SIP30; ZWINT; Zwint-1; ZWINT1
UniProt ID: (Human) O95229, (Rat) Q8VIL3, (Mouse) Q9CQU5
Entrez Gene ID: (Human) 11130, (Rat) 257644, (Mouse) 52696
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.