Novus Biologicals
Manufacturer Code:NBP182869
Catalog # NBP182869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EVRERKTGTQQELDGVFQKLGNLKQQAEQERDKLQRYQTFLQLLYTLQGKLLFPEAEAEAENLPDDKPQQPTRPQEQSTGDTMGRDPGVSFKAVGLQPAGDVN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf164 chromosome 20 open reading frame 164 dJ337O18.7 zinc finger SWIM domain-containing protein 3 zinc finger SWIM domain containing 3 zinc finger SWIM-type containing 3; human ZW10 interacting protein-1; SNAP25 interacting protein of 30 kDa; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; Zwint-1
Gene Aliases: HZwint-1; KNTC2AP; SIP30; ZWINT; ZWINT1
UniProt ID: (Human) O95229
Entrez Gene ID: (Human) 11130
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.