Invitrogen
Manufacturer Code:PA572080
Catalog # PIPA572080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 482-508 amino acids from the C-terminal region of human ZWILCH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of Drosophila Zwilch; hZwilch; KNTC1AP hZwilch homolog of Drosophila Zwilch protein zwilch homolog; Protein zwilch homolog; Zwilch, kinetochore associated, homolog
Gene Aliases: 2310031L18Rik; hZwilch; KNTC1AP; ZWILCH
UniProt ID: (Human) Q9H900, (Mouse) Q8R060
Entrez Gene ID: (Human) 55055, (Mouse) 68014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.