Invitrogen
Manufacturer Code:PA563124
Catalog # PIPA563124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZW10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Centromere/kinetochore protein zw10 homolog; zeste white 10 homolog; ZW10 homolog, centromere/kinetochore protein; ZW10 kinetochore associated homolog (Drosophila) HZW10 KNTC1AP MGC149821; ZW10, kinetochore associated, homolog
Gene Aliases: HZW10; KNTC1AP; ZW10
UniProt ID: (Human) O43264
Entrez Gene ID: (Human) 9183
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.