Invitrogen
Manufacturer Code:PA544885
Catalog # PIPA544885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ZW10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centromere/kinetochore protein zw10; Centromere/kinetochore protein zw10 homolog; zeste white 10 homolog; ZW10 homolog, centromere/kinetochore protein; ZW10 kinetochore associated homolog (Drosophila) HZW10 KNTC1AP MGC149821; ZW10, kinetochore associated, homolog
Gene Aliases: HZW10; KNTC1AP; ZW10
UniProt ID: (Human) O43264, (Rat) Q4V8C2
Entrez Gene ID: (Human) 9183, (Dog) 479433, (Horse) 100062230, (Bovine) 511084, (Rabbit) 100342350, (Guinea pig) 100732304, (Rat) 363059
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.