Invitrogen
Manufacturer Code:PA567602
Catalog # PIPA567602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ZUFSP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf113 dJ412I7.3 zinc finger with UFM1-specific peptidase domain protein; DUB; Lys-63-specific deubiquitinase ZUFSP; Zinc finger with UFM1-specific peptidase domain protein; Zinc finger-containing ubiquitin peptidase 1
Gene Aliases: C6orf113; dJ412I7.3; ZUFSP; ZUP1
UniProt ID: (Human) Q96AP4
Entrez Gene ID: (Human) 221302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.